missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclophilin E Protein
A cDNA sequence encoding the Cyclophilin E was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10572-1mg
Additional Details : Weight : 0.01000kg
Specifications
10450 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 210 nmoles/min/ug, and is defined as the amount of enzyme that cleave 1umole of suc-AAFP-pNA per minute at 1C in Tris-HCl pH8.0 using chymotrypsin. | |
Human | |
His | |
Cyclophilin-E solution containing 20mM Tris pH 8. |
Cyclophilin E Protein | |
Research Use Only | |
PPIE | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMATT KRVLYVGGLA EEVDDKVLHA AFIPFGDITD IQIPLDYETE KHRGFAFVEF ELAEDAAAAI DNMNESELFG RTIRVNLAKP MRIKEGSSRP VWSDDDWLKK FSGKTLEENK EEEGSEPPKA ETQEGEPIAK KARSNPQVYM DIKIGNKPAG RIQMLLRSDV VPMTAENFRC LCTHEKGFGF KGSSFHRIIP QFMCQGGDFT NHNGTGGKSI YGKKFDDENF ILKHTGPGLL SMANSGPNTN GSQFFLTCDK TDWLDGKHVV FGEVTEGLDV LRQIEAQGSK DGKPKQKVII ADCGEYV | |
Greater than 95.0% as determined by SDS-PAGE. |