missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL1 / GRO-alpha Protein
Recombinant Protein
£73.80 - £3020.00
Specifications
Accession Number | NP_001502.1 |
---|---|
Gene ID (Entrez) | 2919 |
Name | CXCL1 / GRO-alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962828
|
enQuireBio™
QP1008-5 UG |
5 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15952828
|
enQuireBio™
QP1008-25 UG |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15942828
|
enQuireBio™
QP1008-100 UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15972828
|
enQuireBio™
QP1008-500 UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15932828
|
enQuireBio™
QP1008-1 MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001502.1 | |
CXCL1 / GRO-alpha Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity confirmed by human neutrophil chemotaxis assay (significanty different from control at Human CXCL1 / GRO-alpha concentrations greater than 10 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
2919 | |
Research Use Only | |
CXCL1 | |
Recombinant Protein | |
E. coli | |
ASVATELRCQ CLQTLQGIHP KNIQSVNVKS PGPHCAQTEV IATLKNGRKA CLNPASPIVK KIIEKMLNSD KSN | |
Greater than 95% as determined by SDS-PAGE. |