missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CTF1 Protein

A cDNA sequence encoding the CTF1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10564-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964258

  • £155.00 / 10µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1489
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0M 106 IU/mg.
Human
Untagged
CTF-1 protein was lyophilized from a 0.2microm filtered concentrated solution in 30% Acetonitrile and 0.1% TFA.
CTF1 Protein
Research Use Only
CTF1
Recombinant Protein
E. coli
MSRREGSLED PQTDSSVSLL PHLEAKIRQT HSLAHLLTKY AEQLLQEYVQ LQGDPFGLPS FSPPRLPVAG LSAPAPSHAG LPVHERLRLD AAALAALPPL LDAVCRRQAE LNPRAPRLLR RLEDAARQAR ALGAAVEALL AALGAANRGP RAEPPAATAS AASATGVFPA KVLGLRVCGL YREWLSRTEG DLGQLLPGGS A
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers