missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSTA Protein
A cDNA sequence encoding the CSTA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11547-GST-2ug
Additional Details : Weight : 0.01000kg
Specifications
1475 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MIPGGLSEAK PATPEIQEIV DKVKPQLEEK TNETYGKLEA VQYKTQVVAG TNYYIKVRAG DNKYMHLKVF KSLPGQNEDL VLTGYQVDKN KDDELTGF |
CSTA Protein | |
Research Use Only | |
CSTA | |
Human | |
GST | |
Cystatin-A in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |