missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CSNK2B Protein

A cDNA sequence encoding the CSNK2B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11534-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947558

  • £5741.17 / 1mg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1460
Greater than 95.0% as determinedAnalysis by SDS-PAGE.
Research Use Only
CSNK2B
Human
Untagged
The CSNK2B protein (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 200mM NaCl, 1mM DTT, 1mM EDTA, 1uM leupeptin and 40% glycerol.
CSNK2B Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSSSEEVSWI SWFCGLRGNE FFCEVDEDYI QDKFNLTGLN EQVPHYRQAL DMILDLEPDE ELEDNPNQSD LIEQAAEMLY GLIHARYILT NRGIAQMLEK YQQGDFGYCP RVYCENQPML PIGLSDIPGE AMVKLYCPKC MDVYTPKSSR HHHTDGAYFG TGFPHMLFMV HPEYRPKRPA NQFVPRLYGF KIHPMAYQLQ LQAASNFKSP VKTIR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers