missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSDC2 Protein
A cDNA sequence encoding the CSDC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11528-2ug
Additional Details : Weight : 0.01000kg
Specifications
CSDC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTSESTS PPVVPPLHSP KSPVWPTFPF HREGSRVWER GGVPPRDLPS PLPTKRTRTY SATARASAGP VFKGVCKQFS RSQGHGFITP ENGSEDIFVH VSDIEGEYVP VEGDEVTYKM CPIPPKNQKF QAVEVVLTQL APHTPHETWS GQVVGS | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CSDC2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |