missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRYBA4 Protein

A cDNA sequence encoding the CRYBA4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11519-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967508

  • £2978.71 / 1mg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CRYBA4 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CRYBA4 protein (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MTLQCTKSAG PWKMVVWDED GFQGRRHEFT AECPSVLELG FETVRSLKVL SGAWVGFEHA GFQGQQYILE RGEYPSWDAW GGNTAYPAER LTSFRPAACA NHRDSRLTIF EQENFLGKKG ELSDDYPSLQ AMGWEGNEVG SFHVHSGAWV CSQFPGYRGF QYVLECDHHS GDYKHFREWG SHAPTFQVQS IRRIQQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers