missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRTAM Protein
A cDNA sequence encoding the CRTAM was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11515-10ug
Additional Details : Weight : 0.01000kg
Specifications
56253 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
SLTNHTETIT VEEGQTLTLK CVTSLRKNSS LQWLTPSGFT IFLNEYPALK NSKYQLLHHS ANQLSITVPN VTLQDEGVYK CLHYSDSVST KEVKVIVLAT PFKPILEASV IRKQNGEEHV VLMCSTMRSK PPPQITWLLG NSMEVSGGTL HEFETDGKKC NTTSTLIIHT YGKNSTVDCI IRHRGLQGRK LVAPFRFEDL VTDEETASDA LERNSLSSQD PQQPTSTVSV TEDSSTSEID KEEKEQTTQD PDLTTEANPQ YLGLARKKSG LEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
CRTAM Protein | |
Research Use Only | |
CRTAM | |
Human | |
His | |
CRTAM protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |