missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRP Protein

A cDNA sequence encoding the CRP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11514-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937498

  • £87.35 / 5µg

Estimated Shipment
05-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1401
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
CRP
Human
Untagged
CRP protein solution (1 mg/ml) containing 20mM Tris-HCl, pH 8.0, 2M Urea and 20% Glycerol.
CRP Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MQTDMSRKAF VFPKESDTSY VSLKAPLTKP LKAFTVCLHF YTELSSTRGY SIFSYATKRQ DNEILIFWSK DIGYSFTVGG SEILFEVPEV TVAPVHICTS WESASGIVEF WVDGKPRVRK SLKKGYTVGA EASIILGQEQ DSFGGNFEGS QSLVGDIGNV NMWDFVLSPD EINTIYLGGP FSPNVLNWRA LKYEVQGEVF TKPQLWP
Videos
SDS
Documents