missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CROT Protein
A DNA sequence encoding the Homo sapiens (Human) CROT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8242-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137407.1 | |
CROT Protein | |
Research Use Only | |
CROT | |
Recombinant Protein | |
E. coli | |
MENQLAKSTE ERTFQYQDSL PSLPVPSLEE SLKKYLESVK PFANQEEYKK TEEIVQKFQS GIGEKLHQKL LERAKGKRNW VFVVIIE | |
Greater than 80% as determined by SDS-PAGE. |
54677 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |