missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRISP2 Protein
A cDNA sequence encoding the CRISP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11506-10ug
Additional Details : Weight : 0.01000kg
Specifications
CRISP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASKDPAFTALLT TQLQVQREIV NKHNELRKAV SPPASNMLKM EWSREVTTNA QRWANKCTLQ HSDPEDRKTS TRCGENLYMS SDPTSWSSAI QSWYDEILDF VYGVGPKSPN AVVGHYTQLV WYSTYQVGCG IAYCPNQDSL KYYYVCQYCP AGNNMNRKNT PYQQGTPCAG CPDDCDKGLC TNSCQYQDLL SNCDSLKNTA GCEHELLKEK CKATCLCENK IY | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CRISP2 filtered (0.4microm) solution at a concentration of 0.3 mg/ml in 50mM acetate buffer, pH 4, 5mM DTT and 20%glycerol. |