missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRISP1 Protein

A cDNA sequence encoding the CRISP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11505-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957468

  • £99.15 / 2µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CRISP1 Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS KKKSARDQFN KLVTDLPNVQ EEIVNIHNAL RRRVVPPASN MLKMSWSEEA AQNARIFSKY CDMTESNPLE RRLPNTFCGE NMHMTSYPVS WSSVIGVWYS ESTSFKHGEW TTTDDDITTD HYTQIVWATS YLIGCAIASC RQQGSPRYLY VCHYCHEGND PETKNEPYKT GVPCEACPSN CEDKLCTNPC IYYDEYFDCD IQVHYLGCNH STTILFCKAT CLCDTEIK
Greater than 85% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein was lyophilized from a 0.5 mg/ml solution containing 20mM TRIS, 50mM NaCl, pH 7.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers