missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COQ9 Protein
A cDNA sequence encoding the COQ9 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | COQ9 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15947418
|
enQuireBio™
QP11488-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937418
|
enQuireBio™
QP11488-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927418
|
enQuireBio™
QP11488-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
COQ9 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
COQ9 protein solution (1 mg/ml) in Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRSSDEQK QQPPNSFSQQ HSETQGAEKP DPESSHSPPR YTDQGGEEEE DYESEEQLQH RILTAALEFV PAHGWTAEAI AEGAQSLGLS SAAASMFGKD GSELILHFVT QCNTRLTRVL EEEQKLVQLG QAEKRKTDQF LRDAVETRLR MLIPYIEHWP RALSILMLPH NIPSSLSLLT SMVDDMWHYA GDQSTDFNWY TRRAMLAAIY NTTELVMMQD SSPDFEDTWR FLENRVNDAM NMGHTAKQVK STGEALVQGL MGAAVTLKNL TGLNQRR | |
Greater than 90.0% as determined by SDS-PAGE. |