missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COMMD9 Protein
A cDNA sequence encoding the COMMD9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11482-1mg
Additional Details : Weight : 0.01000kg
Specifications
COMMD9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAALTAE HFAALQSLLK ASSKDVVRQL CQESFSSSAL GLKKLLDVTC SSLSVTQEEA EELLQALHRL TRLVAFRDLS SAEAILALFP ENFHQNLKNL LTKIILEHVS TWRTEAQANQ ISLPRLVDLD WRVDIKTSSD SISRMAVPTC LLQMKIQEDP SLCGDKPSIS AVTVELSKET LDTMLDGLGR IRDQLSAVAS K | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The COMMD9 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |