missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human COMMD9 Protein

A cDNA sequence encoding the COMMD9 was constructed and used to recombinantly synthesize the protein.

£99.15 - £5730.00

Specifications

Name COMMD9 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15967398
View Documents Promotion Details
enQuireBio™
QP11482-2UG
2 μg
£99.15
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15947398
View Documents Promotion Details
enQuireBio™
QP11482-10UG
10 μg
£155.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15957398
View Documents Promotion Details
enQuireBio™
QP11482-1MG
1 mg
£5730.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

COMMD9 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The COMMD9 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAALTAE HFAALQSLLK ASSKDVVRQL CQESFSSSAL GLKKLLDVTC SSLSVTQEEA EELLQALHRL TRLVAFRDLS SAEAILALFP ENFHQNLKNL LTKIILEHVS TWRTEAQANQ ISLPRLVDLD WRVDIKTSSD SISRMAVPTC LLQMKIQEDP SLCGDKPSIS AVTVELSKET LDTMLDGLGR IRDQLSAVAS K
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications