missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COLEC10 Protein
A DNA sequence encoding the Homo sapiens (Human) COLEC10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8273-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001311024.1 | |
COLEC10 Protein | |
Research Use Only | |
COLEC10 | |
Recombinant Protein | |
E. coli | |
LDIDSRPTAE VCATHTISPG PKGDDGEKGD PGEEGKHGKV GRMGPKGIKG ELGDMGDQGN IGKTGPIGKK GDKGEKGLLG IPGEKGKAGT VCDCGRYRKF VGQLDISIAR LKTSMKFVKN VIAGIRETEE KFYYIVQEEK NYRESLTHCR IRGGMLAMPK DEAANTLIAD YVAKSGFFRV FIGVNDLERE GQYMFTDNTP LQNYSNWNEG EPSDPYGHED CVEMLSSGRW NDTECHLTMY FVCEFIKKKK | |
Greater than 90% as determined by SDS-PAGE. |
10584 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |