missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coagulation factor XIII A chain Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation factor XIII A chain, was expressed in the hosts and tags indicated.
£261.83 - £2888.42
Specifications
Accession Number | NP_000120.2 |
---|---|
Gene ID (Entrez) | 2162 |
Name | Coagulation factor XIII A chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047691
|
enQuireBio™
QP9132-YE-100UG |
100 μg |
£670.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087691
|
enQuireBio™
QP9132-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057691
|
enQuireBio™
QP9132-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077691
|
enQuireBio™
QP9132-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097691
|
enQuireBio™
QP9132-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067691
|
enQuireBio™
QP9132-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000120.2 | |
Coagulation factor XIII A chain Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2162 | |
Research Use Only | |
F13A1 | |
Recombinant Protein | |
Yeast | |
GVNLQEFLNV TSVHLFKERW DTNKVDHHTD KYENNKLIVR RGQSFYVQID FSRPYDPRRD LFRVEYVIGR YPQENKGTYI PVPIVSELQS GKWGAKIVMR EDRSVRLSIQ SSPKCIVGKF RMYVAVWTPY GVLRTSRNPE TDTYILFNPW CEDDAVYLDN EKEREEYVLN DIGVIFYGEV NDIKTRSWSY GQFEDGILDT CLYVMDRAQM DLSGRGNPIK VSRVGSAMVN AKDDEGVLVG SWDNIYAYGV PPSAWTGSVD ILLEYRSSEN PVRYGQCWVF AGVFNTFLRC LGIPARIVTN YFSAHDNDAN LQMDIFLEED GNVNSKLTKD SVWNYHCWNE AWMTRPDLPV GFGGWQAVDS TPQENSDGMY RCGPASVQAI KHGHVCFQFD APFVFAEVNS DLIYITAKKD GTHVVENVDA THIGKLIVTK QIGGDGMMDI TDTYKFQEGQ EEERLALETA LMYGAKKPLN TEGVMKSRSN VDMDFEVENA VLGKDFKLSI TFRNNSHNRY TITAYLSANI TFYTGVPKAE FKKETFDVTL EPLSFKKEAV LIQAGEYMGQ LLEQASLHFF VTARINETRD VLAKQKSTVL TIPEIIIKVR GTQVVGSDMT VTVQFTNPLK ETLRNVWVHL DGPGVTRPMK KMFREIRPNS TVQWEEVCRP WVSGHRKLIA SMSSDSLRHV YGELDVQIQR RPSM | |
Greater than 90% as determined by SDS-PAGE. |