missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNTF Protein
A DNA sequence encoding the Homo sapiens (Human) CNTF, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_000605.1 |
---|---|
Gene ID (Entrez) | 1270 |
Name | CNTF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075121
|
enQuireBio™
QP8530-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015131
|
enQuireBio™
QP8530-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065121
|
enQuireBio™
QP8530-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095121
|
enQuireBio™
QP8530-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005131
|
enQuireBio™
QP8530-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085121
|
enQuireBio™
QP8530-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000605.1 | |
CNTF Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1270 | |
Research Use Only | |
CNTF | |
Recombinant Protein | |
E. coli | |
TEHSPLTPHR RDLCSRSIWL ARKIRSDLTA LTESYVKHQG LNKNINLDSA DGMPVASTDQ WSELTEAERL QENLQAYRTF HVLLARLLED QQVHFTPTEG DFHQAIHTLL LQVAAFAYQI EELMILLEYK IPRNEADGMP INVGDGGLFE KKLWGLKVLQ ELSQWTVRSI HDLRFISSHQ TGIPARGSHY IAN | |
Greater than 90% as determined by SDS-PAGE. |