missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CNDP1 Protein

A cDNA sequence encoding the CNDP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10557-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964238

  • £151.37 / 10µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

84735
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
CNDP1
Recombinant Protein
Insect
SPSPPPALLE KVFQYIDLHQ DEFVQTLKEW VAIESDSVQP VPRFRQELFR MMAVAADTLQ RLGARVASVD MGPQQLPDGQ SLPIPPVILA ELGSDPTKGT VCFYGHLDVQ PADRGDGWLT DPYVLTEVDG KLYGRGATDN KGPVLAWINA VSAFRALEQD LPVNIKFIIE GMEEAGSVAL EELVEKEKDR FFSGVDYIVI SDNLWISQRK PAITYGTRGN SYFMVEVKCR DQDFHSGTFG GILHEPMADL VALLGSLVDS SGHILVPGIY DEVVPLTEEE INTYKAIHLD LEEYRNSSRV EKFLFDTKEE ILMHLWRYPS LSIHGIEGAF DEPGTKTVIP GRVIGKFSIR LVPHMNVSAV EKQVTRHLED VFSKRNSSNK MVVSMTLGLH PWIANIDDTQ YLAAKRAIRT VFGTEPDMIR DGSTIPIAKM FQEIVHKSVV LIPLGAVDDG EHSQNEKINR WNYIEGTKLF AAFFLEMAQL HLEHHHHHH
CNDP1 Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is >3,000 pmol/min/ug, and as measured by the hydrolysis of carnosine per minute at pH6.8 at 25C.
Human
His
CNDP1 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers