missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLPS Protein
A cDNA sequence encoding the CLPS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11452-20ug
Additional Details : Weight : 0.01000kg
Specifications
CLPS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAPGPRGI IINLENGELC MNSAQCKSNC CQHSSALGLA RCTSMASENS ECSVKTLYGI YYKCPCERGL TCEGDKTIVG SITNTNFGIC HDAGRSKQ | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CLPS solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 1mM DTT and 0.1M NaCl. |