missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CLIC4 Protein

A DNA sequence encoding the Homo sapiens (Human) CLIC4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5838-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958129

  • £384.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_039234.1
CLIC4 Protein
Research Use Only
CLIC4
Recombinant Protein
E. coli
MALSMPLNGL KEEDKEPLIE LFVKAGSDGE SIGNCPFSQR LFMILWLKGV VFSVTTVDLK RKPADLQNLA PGTHPPFITF NSEVKTDVNK IEEFLEEVLC PPKYLKLSPK HPESNTAGMD IFAKFSAYIK NSRPEANEAL ERGLLKTLQK LDEYLNSPLP DEIDENSMED IKFSTRKFLD GNEMTLADCN LLPKLHIVKV VAKKYRNFDI PKEMTGIWRY LTNAYSRDEF TNTCPSDKEV EIAYSDVAKR LTK
Greater than 80% as determined by SDS-PAGE.
25932
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers