missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLIC4 Protein
A DNA sequence encoding the Homo sapiens (Human) CLIC4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5838-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_039234.1 | |
CLIC4 Protein | |
Research Use Only | |
CLIC4 | |
Recombinant Protein | |
E. coli | |
MALSMPLNGL KEEDKEPLIE LFVKAGSDGE SIGNCPFSQR LFMILWLKGV VFSVTTVDLK RKPADLQNLA PGTHPPFITF NSEVKTDVNK IEEFLEEVLC PPKYLKLSPK HPESNTAGMD IFAKFSAYIK NSRPEANEAL ERGLLKTLQK LDEYLNSPLP DEIDENSMED IKFSTRKFLD GNEMTLADCN LLPKLHIVKV VAKKYRNFDI PKEMTGIWRY LTNAYSRDEF TNTCPSDKEV EIAYSDVAKR LTK | |
Greater than 80% as determined by SDS-PAGE. |
25932 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |