missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Clathrin light chain A Protein
A DNA sequence encoding the Homo sapiens (Human) Clathrin light chain A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5840-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001070145.1 | |
Clathrin light chain A Protein | |
Research Use Only | |
Clta | |
Recombinant Protein | |
E. coli | |
MAELDPFGAP AGAPGGPALG NGVAGAGEED PAAAFLAQQE SEIAGIENDE AFAILDGGAP GPQPHGEPPG GPDAVDGVMN GEYYQESNGP TDSYAAISQV DRLQSEPESI RKWREEQMER LEALDANSRK QEAEWKEKAI KELEEWYARQ DEQLQKTKAN NRAAEEAFVN DIDESSPGTE WERVARLCDF NPKSSKQAKD VSRMRSVLIS LKQAPLVH | |
Greater than 80% as determined by SDS-PAGE. |
1211 | |
200 μg | |
Not determined. | |
Full Length of Isoform Non-brain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |