missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CK2h Protein
A cDNA sequence encoding the CK2h was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11434-10ug
Additional Details : Weight : 0.01000kg
Specifications
CK2h Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
CK2 Holoenzyme is supplied 0.5 mg/1ml in 25mM Tris-HCl, 500mM NaCl, 1mM DTT, 500 microm PMSF, 5% glycerol, pH 8.5. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 95% as determined by SDS-PAGE. |