missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CK2h Protein
A cDNA sequence encoding the CK2h was constructed and used to recombinantly synthesize the protein.
£439.00 - £1655.00
Specifications
Name | CK2h Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987248
|
enQuireBio™
QP11434-3UG |
3 μg |
£439.00
3µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967248
|
enQuireBio™
QP11434-10UG |
10 μg |
£880.00
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977248
|
enQuireBio™
QP11434-20UG |
20 μg |
£1655.00
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CK2h Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 95% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
CK2 Holoenzyme is supplied 0.5 mg/1ml in 25mM Tris-HCl, 500mM NaCl, 1mM DTT, 500 microm PMSF, 5% glycerol, pH 8.5. |