missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cilia- and flagella-associated protein 20 Protein
A DNA sequence encoding the Homo sapiens (Human) Cilia- and flagella-associated protein 20, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_037374.1 |
---|---|
Gene ID (Entrez) | 29105 |
Name | Cilia- and flagella-associated protein 20 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073761
|
enQuireBio™
QP8284-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093761
|
enQuireBio™
QP8284-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083761
|
enQuireBio™
QP8284-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003771
|
enQuireBio™
QP8284-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013771
|
enQuireBio™
QP8284-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023771
|
enQuireBio™
QP8284-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_037374.1 | |
Cilia- and flagella-associated protein 20 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
29105 | |
Research Use Only | |
C16orf80 | |
Recombinant Protein | |
E. coli | |
MFKNTFQSGF LSILYSIGSK PLQIWDKKVR NGHIKRITDN DIQSLVLEIE GTNVSTTYIT CPADPKKTLG IKLPFLVMII KNLKKYFTFE VQVLDDKNVR RRFRASNYQS TTRVKPFICT MPMRLDDGWN QIQFNLLDFT RRAYGTNYIE TLRVQIHANC RIRRVYFSDR LYSEDELPAE FKLYLPVQNK AKQ | |
Greater than 80% as determined by SDS-PAGE. |