missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CIAO1 Protein
A DNA sequence encoding the Homo sapiens (Human) CIAO1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5836-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004795.1 | |
CIAO1 Protein | |
Research Use Only | |
CIAO1 | |
Recombinant Protein | |
E. coli | |
MKDSLVLLGR VPAHPDSRCW FLAWNPAGTL LASCGGDRRI RIWGTEGDSW ICKSVLSEGH QRTVRKVAWS PCGNYLASAS FDATTCIWKK NQDDFECVTT LEGHENEVKS VAWAPSGNLL ATCSRDKSVW VWEVDEEDEY ECVSVLNSHT QDVKHVVWHP SQELLASASY DDTVKLYREE EDDWVCCATL EGHESTVWSL AFDPSGQRLA SCSDDRTVRI WRQYLPGNEQ GVACSGSDPS WKCICTLSGF HSRTIYDIAW CQLTGALATA CGDDAIRVFQ EDPNSDPQQP TFSLTAHLHQ AHSQDVNCVA WNPKEPGLLA SCSDDGEVAF WKYQRPEGL | |
Greater than 90% as determined by SDS-PAGE. |
9391 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |