missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Chromogranin A Protein

A DNA sequence encoding the Homo sapiens (Human) Chromogranin A, was expressed in the hosts and tags indicated.

£183.90 - £1963.71

Specifications

Name Chromogranin A Protein
Product Type Recombinant Protein
Protein Tag His
Host Species Yeast
Cross Reactivity Human
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15938059
View Documents
enQuireBio™
QP5827-YE-100UG
100 μg
£405.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment
01-06-2021
Please to purchase this item. Need a web account? Register with us today!
15978059
View Documents
enQuireBio™
QP5827-YE-500UG
500 μg
£1115.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment
01-06-2021
Please to purchase this item. Need a web account? Register with us today!
15948059
View Documents
enQuireBio™
QP5827-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15968059
View Documents
enQuireBio™
QP5827-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15988059
View Documents
enQuireBio™
QP5827-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15958059
View Documents
enQuireBio™
QP5827-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

Chromogranin A Protein
His
Human
Greater than 90% as determined by SDS-PAGE.
NP_001266.1
Recombinant Protein
Yeast
LPVNSPMNKG DTEVMKCIVE VISDTLSKPS PMPVSQECFE TLRGDERILS ILRHQNLLKE LQDLALQGAK ERAHQQKKHS GFEDELSEVL ENQSSQAELK EAVEEPSSKD VMEKREDSKE AEKSGEATDG ARPQALPEPM QESKAEGNNQ APGEEEEEEE EATNTHPPAS LPSQKYPGPQ AEGDSEGLSQ GLVDREKGLS AEPGWQAKRE EEEEEEEEAE AGEEAVPEEE GPTVVLNPHP SLGYKEIRKG ESRSEALAVD GAGKPGAEEA QDPEGKGEQE HSQQKEEEEE MAVVPQGLFR GGKSGELEQE EERLSKEWED SKRWSKMDQL AKELTAEKRL EGQEEEEDNR DSSMKLSFRA RAYGFRGPGP QLRRGWRPSS REDSLEAGLP LQVRGYPEEK KEEEGSANRR PEDQELESLS AIEAELEKVA HQLQALRRG
1113
CHGA
SDS