missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Chromodomain-helicase-DNA-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Chromodomain-helicase-DNA-binding protein 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8347-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001264.2 | |
Chromodomain-helicase-DNA-binding protein 1 | |
Research Use Only | |
CHD4 | |
Recombinant Protein | |
E. coli | |
MASGLGSPSP CSAGSEEEDM DALLNNSLPP PHPENEEDPE EDLSETETPK LKKKKKPKKP RDPKIPKSKR QKKERMLLCR QLGDSSGEGP EFVEEEEEVA LRSDSEGSDY TPGKKKKKKL GPKKEKKSKS KRKEEEEEED DDDDSKEPKS SAQLLEDWGM EDIDHVFSEE DYRTLTNYKA FSQFVRPLIA AKNPKIAVSK MMMVLGAKWR EFSTNNPFK | |
Greater than 90% as determined by SDS-PAGE. |
1108 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |