missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CHMP1A Protein

A cDNA sequence encoding the CHMP1A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11420-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957208

  • £155.00 / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

5119
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDDTLFQ LKFTAKQLEK LAKKAEKDSK AEQAKVKKAL LQKNVECARV YAENAIRKKN EGVNWLRMAS RVDAVASKVQ TAVTMKGVTK NMAQVTKALD KALSTMDLQK VSSVMDRFEQ QVQNLDVHTS VMEDSMSSAT TLTTPQEQVD SLIMQIAEEN GLEVLDQLSQ LPEGASAVGE SSVRSQEDQL SRRLAALRN
Greater than 85.0% as determined by SDS-PAGE.
CHMP1A Protein
Research Use Only
CHMP1A
Human
His
CHMP1A protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers