missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHMP1A Protein
A cDNA sequence encoding the CHMP1A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11420-100ug
Additional Details : Weight : 0.01000kg
Specifications
5119 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDDTLFQ LKFTAKQLEK LAKKAEKDSK AEQAKVKKAL LQKNVECARV YAENAIRKKN EGVNWLRMAS RVDAVASKVQ TAVTMKGVTK NMAQVTKALD KALSTMDLQK VSSVMDRFEQ QVQNLDVHTS VMEDSMSSAT TLTTPQEQVD SLIMQIAEEN GLEVLDQLSQ LPEGASAVGE SSVRSQEDQL SRRLAALRN | |
Greater than 85.0% as determined by SDS-PAGE. |
CHMP1A Protein | |
Research Use Only | |
CHMP1A | |
Human | |
His | |
CHMP1A protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 20% glycerol and 1mM DTT. |