missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHAC2 Protein
A cDNA sequence encoding the CHAC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11401-10ug
Additional Details : Weight : 0.01000kg
Specifications
CHAC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMWVFGYG SLIWKVDFPY QDKLVGYITN YSRRFWQGST DHRGVPGKPG RVVTLVEDPA GCVWGVAYRL PVGKEEEVKA YLDFREKGGY RTTTVIFYPK DPTTKPFSVL LYIGTCDNPD YLGPAPLEDI AEQIFNAAGP SGRNTEYLFE LANSIRNLVP EEADEHLFAL EKLVKERLEG KQNLNCI | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CHAC2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl and 10% glycerol. |