missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CGB Protein

A cDNA sequence encoding the CGB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11398-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957138

  • £2978.71 / 1mg
Estimated Shipment: 04-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CGB Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CGB protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSKEPLRP RCRPINATLA VEKEGCPVCI TVNTTICAGY CPTMTRVLQG VLPALPQVVC NYRDVRFESI RLPGCPRGVN PVVSYAVALS CQCALCRRST TDCGGPKDHP LTCDDPRFQD SSSSKAPPPS LPSPSRLPGP SDTPILPQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers