missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Centromere protein 1

A DNA sequence encoding the Homo sapiens (Human) Centromere protein K, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5819-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947989

  • £560.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001253967.1
Centromere protein 1
Research Use Only
CENPK
Recombinant Protein
E. coli
MNQEDLDPDS TTDVGDVTNT EEELIRECEE MWKDMEECQN KLSLIGTETL TDSNAQLSLL IMQVKCLTAE LSQWQKKTPE TIPLTEDVLI TLGKEEFQKL RQDLEMVLST KESKNEKLKE DLEREQRWLD EQQQIMESLN VLHSELKNKV ETFSESRIFN ELKTKMLNIK EYKEKLLSTL GEFLEDHFPL PDRSVKKKKK NIQESSVNLI TLHEMLEILI NRLFDVPHDP YVKISDSFWP PYVELLLRNG IALRHPEDPT RIRLEAFHQ
Greater than 80% as determined by SDS-PAGE.
64105
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers