missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CEL Protein

A cDNA sequence encoding the CEL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11385-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967098

  • £1320.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1056
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human
AKLGAVYTEG GFVEGVNKKL GLLGDSVDIF KGIPFAAPTK ALENPQPHPG WQGTLKAKNF KKRCLQATIT QDSTYGDEDC LYLNIWVPQG RKQVSRDLPV MIWIYGGAFL MGSGHGANFL NNYLYDGEEI ATRGNVIVVT FNYRVGPLGF LSTGDANLPG NYGLRDQHMA IAWVKRNIAA FGGDPNNITL FGESAGGASV SLQTLSPYNK GLIRRAISQS GVALSPWVIQ KNPLFWAKKV AEKVGCPVGD AARMAQCLKV TDPRALTLAY KVPLAGLEYP MLHYVGFVPV IDGDFIPADP INLYANAADI DYIAGTNNMD GHIFASIDMP AINKGNKKVT EEDFYKLVSE FTITKGLRGA KTTFDVYTES WAQDPSQENK KKTVVDFETD VLFLVPTEIA LAQHRANAKS AKTYAYLFSH PSRMPVYPKW VGADHADDIQ YVFGKPFATP TGYRPQDRTV SKAMIAYWTN FAKTGDPNMG DSAVPTHWEP YTTENSGYLE ITKKMGSSSM KRSLRTNFLR YWTLTYLALP TVTDQEATPV PPTGDSEATP VPPTGDSETA PVPPTGDSGA PPVPPTGDSG APPVPPTGDS GAPPVPPTGD SGAPPVPPTG DSGAPPVPPT GDSGAPPVPP TGDSGAPPVP PTGDSGAPPV PPTGDAGPPP VPPTGDSGAP PVPPTGDSGA PPVTPTGDSE TAPVPPTGDS GAPPVPPTGD SEAAPVPPTD DSKEAQMPAV IRF
Greater than 95.0% as determined by SDS-PAGE.
CEL Protein
Research Use Only
CEL
Human
Untagged
CEL protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers