missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDKN3 Protein
A cDNA sequence encoding the CDKN3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11379-1mg
Additional Details : Weight : 0.01000kg
Specifications
1033 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKPPSSIQTS EFDSSDEEPI EDEQTPIHIS WLSLSRVNCS QFLGLCALPG CKFKDVRRNV QKDTEELKSC GIQDIFVFCT RGELSKYRVP NLLDLYQQCG IITHHHPIAD GGTPDIASCC EIMEELTTCL KNYRKTLIHC YGGLGRSCLV AACLLLYLSD TISPEQAIDS LRDLRGSGAI QTIKQYNYLH EFRDKLAAHL SSRDSQSRSV SR | |
Greater than 90.0% as determined by SDS-PAGE. |
CDKN3 Protein | |
Research Use Only | |
CDKN3 | |
Human | |
His | |
The CDKN3 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 40% glycerol and 0.1M NaCl. |