missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDKN2D / p19ink4d Protein

A DNA sequence encoding the Homo sapiens (Human) CDKN2D / p19ink4d, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5809-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967909

  • £560.06 / 100µg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001791.1
CDKN2D / p19ink4d Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
1032
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CDKN2D
Recombinant Protein
E. coli
MLLEEVRAGD RLSGAAARGD VQEVRRLLHR ELVHPDALNR FGKTALQVMM FGSTAIALEL LKQGASPNVQ DTSGTSPVHD AARTGFLDTL KVLVEHGADV NVPDGTGALP IHLAVQEGHT AVVSFLAAES DLHRRDARGL TPLELALQRG AQDLVDILQG HMVAPL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers