missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK3 Protein
A cDNA sequence encoding the CDK3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11375-2ug
Additional Details : Weight : 0.01000kg
Specifications
1018 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDMFQKV EKIGEGTYGV VYKAKNRETG QLVALKKIRL DLEMEGVPST AIREISLLKE LKHPNIVRLL DVVHNERKLY LVFEFLSQDL KKYMDSTPGS ELPLHLIKSY LFQLLQGVSF CHSHRVIHRD LKPQNLLINE LGAIKLADFG LARAFGVPLR TYTHEVVTLW YRAPEILLGS KFYTTAVDIW SIGCIFAEMV TRKALFPGDS EIDQLFRIFR MLGTPSEDTW PGVTQLPDYK GSFPKWTRKG LEEIVPNLEP EGRDLLMQLL QYDPSQRITA KTALAHPYFS SPEPSPAARQ YVLQRFRH | |
Greater than 85% as determined by SDS-PAGE. |
CDK3 Protein | |
Research Use Only | |
CDK3 | |
Human | |
His | |
The CDK3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 10% glycerol. |