missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK2AP1 Protein
A DNA sequence encoding the Homo sapiens (Human) CDK2AP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5805-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001257362.1 | |
CDK2AP1 Protein | |
Research Use Only | |
CDK2AP1 | |
Recombinant Protein | |
E. coli | |
MSYKPNLAAH MPAAALNAAG SVHSPSTSMA TSSQYRQLLS DYGPPSLGYT QGTGNSQVPQ SKYAELLAII EELGKEIRPT YAGSKSAMER LKRGIIHARG LVRECLAETE RNARS | |
Greater than 90% as determined by SDS-PAGE. |
8099 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |