missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDCA8 Protein
A cDNA sequence encoding the CDCA8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11366-1mg
Additional Details : Weight : 0.01000kg
Specifications
CDCA8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRKLASFLKD FDREVEIRIK QIESDRQNLL KEVDNLYNIE ILRLPKALRE MNWLDYFALG GNKQALEEAA TADLDITEIN KLTAEAIQTP LKSAKTRKVI QVDEMIVEEE EEEENERKNL QTARVKRCPP SKKRTQSIQG KGKGKRSSRA NTVTPAVGRLEVSMVKPTPG LTPRFDSRVF KTPGLRTPAA GERIYNISGN GSPLADSKEI FLTVPVGGGE SLRLLASDLQ RHSIAQLDPE ALGNIKKLSN RLAQICSSIR THK | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CDCA8 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 1mM DTT, 1mM EDTA and 0.2M NaCl. |