missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD93 Protein
A cDNA sequence encoding the CD93 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11357-1ug
Additional Details : Weight : 0.01000kg
Specifications
CD93 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
TGADTEAVVC VGTACYTAHS GKLSAAEAQN HCNQNGGNLA TVKSKEEAQH VQRVLAQLLR REAALTARMS KFWIGLQREK GKCLDPSLPL KGFSWVGGGE DTPYSNWHKE LRNSCISKRC VSLLLDLSQP LLPSRLPKWS EGPCGSPGSP GSNIEGFVCK FSFKGMCRPL ALGGPGQVTY TTPFQTTSSS LEAVPFASAA NVACGEGDKD ETQSHYFLCK EKAPDVFDWG SSGPLCVSPK YGCNFNNGGC HQDCFEGGDG SFLCGCRPGF RLLDDLVTCA SRNPCSSSPC RGGATCVLGP HGKNYTCRCP QGYQLDSSQL DCVDVDECQD SPCAQECVNT PGGFRCECWV GYEPGGPGEG ACQDVDECAL GRSPCAQGCT NTDGSFHCSC EEGYVLAGED GTQCQDVDEC VGPGGPLCDS LCFNTQGSFH CGCLPGWVLA PNGVSCTMGP VSLGPPSGPP DEEDKGEKEG STVPRAATAS PTRGPEGTPK ATPTTSRPSL SSDAPITSAP LKMLAPSGSP GVWREPSIHH ATAASGPQEP AGGDSSVATQ NNDGTDGQKV EHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CD93 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |