missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD9 Protein
A cDNA sequence encoding the CD9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11354-1mg
Additional Details : Weight : 0.01000kg
Specifications
928 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSHKDEVI KEVQEFYKDT YNKLKTKDEP QRETLKAIHY ALNCCGLAGG VEQFISDICP KKDVLETFTV KSCPDAIKEV FDNKFHI | |
Greater than 85.0% as determined by SDS-PAGE. |
CD9 Protein | |
Research Use Only | |
CD9 | |
Human | |
His | |
The CD9 solution (0.25 mg/1ml) contains Phosphate buffer saline (pH 7.4), 20% glycerol and 1mM DTT. |