missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD84 Protein

A cDNA sequence encoding the CD84 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11352-HIS-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956978

  • £155.00 / 20µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

8832
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMKDSEI FTVNGILGES VTFPVNIQEP RQVKIIAWTS KTSVAYVTPG DSETAPVVTV THRNYYERIH ALGPNYNLVI SDLRMEDAGD YKADINTQAD PYTTTKRYNL QIYRRLGKPK ITQSLMASVN STCNVTLTCS VEKEEKNVTY NWSPLGEEGN VLQIFQTPED QELTYTCTAQ NPVSNNSDSI SARQLCADIA MGFRTHHTG
Greater than 80% as determined by SDS-PAGE.
CD84 Protein
Research Use Only
CD84
Human
His
The CD84 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers