missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD7 Protein
A DNA sequence encoding the Homo sapiens (Human) CD7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5794-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006128.1 | |
CD7 Protein | |
Research Use Only | |
CD7 | |
Recombinant Protein | |
E. coli | |
AQEVQQSPHC TTVPVGASVN ITCSTSGGLR GIYLRQLGPQ PQDIIYYEDG VVPTTDRRFR GRIDFSGSQD NLTITMHRLQ LSDTGTYTCQ AITEVNVYGS GTLVLVTEEQ SQGWHRCSDA PPRASALPAP PTGSALPDPQ TASALPDPPA ASALP | |
Greater than 90% as determined by SDS-PAGE. |
924 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |