missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD58 Protein

A cDNA sequence encoding the CD58 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11340-HIS-EC-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966928

  • £2978.71 / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

965
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
CD58
Human
His
CD58 protein solution (1 mg/ml) containing 20mM Phosphate buffer and 10% glycerol.
CD58 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSFSQQIYG VVYGNVTFHV PSNVPLKEVL WKKQKDKVAE LENSEFRAFS SFKNRVYLDT VSGSLTIYNL TSSDEDEYEM ESPNITDTMK FFLYVLESLP SPTLTCALTN GSIEVQCMIP EHYNSHRGLI MYSWDCPMEQ CKRNSTSIYF KMENDLPQKI QCTLSNPLFN TTSSIILTTC IPSSGHSRHR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers