missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD5 Protein
A cDNA sequence encoding the CD5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11338-HIS-20ug
Additional Details : Weight : 0.01000kg
Specifications
921 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRLSWYDP DFQARLTRSN SKCQGQLEVY LKDGWHMVCS QSWGRSSKQW EDPSQASKVC QRLNCGVPLS LGPFLVTYTP QSSIICYGQL GSFSNCSHSR NDMCHSLGLT CLEPQKTTPP TTRPPPTTTP EPTAPPRLQL VAQSGGQHCA GVVEFYSGSL GGTISYEAQD KTQDLENFLC NNLQCGSFLK HLPETEAGRA QDPGEPREHQ PLPIQWKIQN SSCTSLEHCF RKIKPQKSGR VLALLCSGFQ PKVQSRLVGG SSICEGTVEV RQGAQWAALC DSSSARSSLR WEEVCREQQC GSVNSYRVLD AGDPTSRGLF CPHQKLSQCH ELWERNSYCK KVFVTCQDPN P | |
Greater than 80% as determined by SDS-PAGE. |
CD5 Protein | |
Research Use Only | |
CD5 | |
Human | |
His | |
The CD5 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |