missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD3G Protein
A cDNA sequence encoding the CD3G was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11332-HIS-1mg
Additional Details : Weight : 0.01000kg
Specifications
917 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSQSIKGNH LVKVYDYQED GSVLLTCDAE AKNITWFKDG KMIGFLTEDK KKWNLGSNAK DPRGMYQCKG SQNKSKPLQV YYRMCQNCIE LNAATIS | |
Greater than 90% as determined by SDS-PAGE. |
CD3G Protein | |
Research Use Only | |
CD3G | |
Human | |
His | |
CD3G protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |