missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD37 Protein

A cDNA sequence encoding the CD37 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11330-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946888

  • £155.00 / 20µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

951
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQRAQLER SLRDVVEKTI QKYGTNPEET AAEESWDYVQ FQLRCCGWHY PQDWFQVLIL RGNGSEAHRV PCSCYNLSAT NDSTILDKVI LPQLSRLGHL ARSRHSADIC AVPAESHIYR EGCAQGLQKW LHNN
Greater than 90.0% as determined by SDS-PAGE.
CD37 Protein
Research Use Only
CD37
Human
His
CD37 protein solution (1 mg/ml) containing Phosphate buffer saline, (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers