missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD37 Protein
A cDNA sequence encoding the CD37 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11330-5ug
Additional Details : Weight : 0.01000kg
Specifications
951 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQRAQLER SLRDVVEKTI QKYGTNPEET AAEESWDYVQ FQLRCCGWHY PQDWFQVLIL RGNGSEAHRV PCSCYNLSAT NDSTILDKVI LPQLSRLGHL ARSRHSADIC AVPAESHIYR EGCAQGLQKW LHNN | |
Greater than 90.0% as determined by SDS-PAGE. |
CD37 Protein | |
Research Use Only | |
CD37 | |
Human | |
His | |
CD37 protein solution (1 mg/ml) containing Phosphate buffer saline, (pH 7.4) and 10% glycerol. |