missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD34 Protein

A cDNA sequence encoding the CD34 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11328-HIS-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946878

  • £151.37 / 10µg
Estimated Shipment: 01-03-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

947
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MLVRRGARAG PRMPRGWTAL CLLSLLPSGF MSLDNNGTAT PELPTQGTFS NVSTNVSYQE TTTPSTLGST SLHPVSQHGN EATTNITETT VKFTSTSVIT SVYGNTNSSV QSQTSVISTV FTTPANVSTP ETTLKPSLSP GNVSDLSTTS TSLATSPTKP YTSSSPILSD IKAEIKCSGI REVKLTQGIC LEQNKTSSCA EFKKDRGEGL ARVLCGEEQA DADAGAQVCS LLLAQSEVRP QCLLLVLANR TEISSKLQLM KKHQSDLKKL GILDFTEQDV ASHQSYSQKT LIALVTSGAL LAVLGITGYF LMNRRSWSPT GERLGEDPYY TENGGGQGYS SGPGTSPEAQ GKASVNRGAQ ENGTGQATSR NGHSARQHVV ADTEL
CD34 Protein
Research Use Only
CD34
Human
His
CD34 in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced).
Videos
SDS
Documents
Certificates
Special Offers

Special Offers